Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

enkurin Antibody (OTI6C10), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP272461IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272461IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272461IR Supplier Novus Biologicals Supplier No. NBP272461IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

enkurin Monoclonal specifically detects enkurin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen enkurin
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6C10
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C10orf63DKFZp781F21103, chromosome 10 open reading frame 63, enkurin, enkurin, TRPC channel interacting protein, MGC26778
Gene Symbols ENKUR
Host Species Mouse
Immunogen Full length recombinant protein of human enkurin (NP_659447) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 219670.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.