Learn More
Description
Ensure accurate, reproducible results in Western Blot
Specifications
Specifications
| Antigen | Enolase 1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 2217 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:5000-1:20000 |
| Formulation | PBS with No Preservative |
| Gene Alias | 2-phospho-D-glycerate hydro-lyase, Alpha enolase, C-myc promoter-binding protein, c-myc promoter-binding protein-1, EC 4.2.1, EC 4.2.1.11, ENO1L1, Enolase 1, enolase 1, (alpha), MBP-1, MBPB1, MPB-1, MPB1c-myc promoter-binding protein 1, MYC promoter-binding protein 1, NNE, Non-neural enolase, Phosphopyruvate hydratase, Plasminogen-binding protein, PPHalpha enolase like 1, tau-crystallin |
| Gene Symbols | ENO1 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
