Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enolase 1 Antibody (253), Novus Biologicals™
SDP

Catalog No. NBP225147SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225147SS 0.025 mL
NBP225147 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP225147SS Supplier Novus Biologicals Supplier No. NBP225147SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Enolase 1 Monoclonal specifically detects Enolase 1 in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay.

Specifications

Antigen Enolase 1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay
Classification Monoclonal
Clone 253
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:10000, Simple Western 1:500, Immunohistochemistry 1:2000-1:5000, Immunocytochemistry/Immunofluorescence 1:2000-1:5000, Immunoassay
Formulation 50% PBS, 50% glycerol with 5mM Sodium Azide
Gene Accession No. P06733
Gene Alias 2-phospho-D-glycerate hydro-lyase, Alpha enolase, C-myc promoter-binding protein, c-myc promoter-binding protein-1, EC 4.2.1, EC 4.2.1.11, ENO1L1, Enolase 1, enolase 1, (alpha), MBP-1, MBPB1, MPB-1, MPB1c-myc promoter-binding protein 1, MYC promoter-binding protein 1, NNE, Non-neural enolase, Phosphopyruvate hydratase, Plasminogen-binding protein, PPHalpha enolase like 1, tau-crystallin
Gene Symbols ENO1
Host Species Mouse
Immunogen The N-terminal 12 amino acids of bovine Enolase 1, which was synthesized on a 8-amine lysine core. [UniProt# Q9XSJ4]
Molecular Weight of Antigen 47 kDa
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2023
Test Specificity Human, Bovine, Mouse, Rat, Xenopus. Other species have not been tested.
Target Species Human, Mouse, Rat, Pig, Bovine, Equine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.