Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enolase 2/Neuron-specific Enolase Antibody, Novus Biologicals™
SDP

Catalog No. NB1001606 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001606 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001606 Supplier Novus Biologicals Supplier No. NB1001606
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Enolase 2/Neuron-specific Enolase Polyclonal specifically detects Enolase 2/Neuron-specific Enolase in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Western Blot.

Specifications

Antigen Enolase 2/Neuron-specific Enolase
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:2000-1:5000, Immunohistochemistry-Paraffin 1:2000-1:5000
Formulation 10mM Sodium PBS (0.9% isotonic, w/v, pH 7.2) with 0.02% Sodium Azide
Gene Accession No. P09104
Gene Alias 2-phospho-D-glycerate hydrolyase, 2-phospho-D-glycerate hydro-lyase, EC 4.2.1.11, Enolase 2, enolase 2 (gamma, neuronal), gamma-enolase, Neural enolase, neuron specific gamma enolase, neurone-specific enolase, Neuron-specific enolase, NSE
Gene Symbols ENO2
Host Species Chicken
Immunogen Chickens were immunized with two synthetic peptide/keyhole limpet hemocyanin (KLH) conjugates. These synthetic peptides corresponded to different regions of the NSE-2 gene product, but are shared between the human (NP_001966, NCBI) and rat (AAA41119, NCBI) sequences.
Purification Method Affinity Purified
Quantity 0.25 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2026
Test Specificity Neuron-Specific Enolase (type 2)
Target Species Human, Mouse, Rat, Rat
Content And Storage Store at 4C in the dark.
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.