Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enolase 2/Neuron-specific Enolase Antibody (NSEP1), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP250532C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250532C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250532C Supplier Novus Biologicals Supplier No. NBP250532C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Enolase 2/Neuron-specific Enolase Monoclonal specifically detects Enolase 2/Neuron-specific Enolase in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen Enolase 2/Neuron-specific Enolase
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone NSEP1
Conjugate DyLight 650
Dilution Western Blot, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 2-phospho-D-glycerate hydrolyase, 2-phospho-D-glycerate hydro-lyase, EC 4.2.1.11, Enolase 2, enolase 2 (gamma, neuronal), gamma-enolase, Neural enolase, neuron specific gamma enolase, neurone-specific enolase, Neuron-specific enolase, NSE
Gene Symbols ENO2
Host Species Mouse
Immunogen Ovalbumin-conjugated synthetic peptides corresponding to human NSE amino acid sequence:NSE-P1: aa's 416-433 - LGDEARFAGHNFRNPSVL
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2026
Test Specificity Not reactive with -isozyme of enolase.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.