Learn More
Enolase 2/Neuron-specific Enolase Antibody (SPM347), Allophycocyanin/Cy7, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Enolase 2/Neuron-specific Enolase |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | SPM347 |
| Conjugate | APC-Cyanine7 |
| Dilution | Flow Cytometry |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | 2-phospho-D-glycerate hydrolyase, 2-phospho-D-glycerate hydro-lyase, EC 4.2.1.11, Enolase 2, enolase 2 (gamma, neuronal), gamma-enolase, Neural enolase, neuron specific gamma enolase, neurone-specific enolase, Neuron-specific enolase, NSE |
| Gene Symbols | ENO2 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.