Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enolase 2/Neuron-specific Enolase Antibody (SPM347) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB254590200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB254590200 200 μg
NB254590100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB254590200 Supplier Novus Biologicals Supplier No. NBP254590200UG

Mouse Monoclonal Antibody

Enolase 2/Neuron-specific Enolase Monoclonal antibody specifically detects Enolase 2/Neuron-specific Enolase in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.

Specifications

Antigen Enolase 2/Neuron-specific Enolase
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone SPM347
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.1 - 0.2 ug/ml, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias 2-phospho-D-glycerate hydrolyase, 2-phospho-D-glycerate hydro-lyase, EC 4.2.1.11, Enolase 2, enolase 2 (gamma, neuronal), gamma-enolase, Neural enolase, neuron specific gamma enolase, neurone-specific enolase, Neuron-specific enolase, NSE
Gene Symbols ENO2
Host Species Mouse
Immunogen A synthetic peptide corresponding to aa416-433 of human NSE gamma (exact sequence is proprietary)
Molecular Weight of Antigen 50 kDa
Purification Method Protein A or G purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2026
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.