Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eNOS Antibody (6H2), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP151582C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP151582C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP151582C Supplier Novus Biologicals Supplier No. NBP151582C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

eNOS Monoclonal specifically detects eNOS in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen eNOS
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6H2
Conjugate DyLight 650
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Constitutive NOS, EC 1.14.13.39, ECNOS, endothelial, Endothelial NOS, eNOScNOS, nitric oxide synthase 3 (endothelial cell), NOS type III, NOSIII
Gene Symbols NOS3
Host Species Mouse
Immunogen Purified recombinant fragment of human eNOS expressed in E. coli. [UniProt# P29474]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4846
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.