Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eNOS Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB3050AF532 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB3050AF532 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB3050AF532 Supplier Novus Biologicals Supplier No. NB300500AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

eNOS Polyclonal specifically detects eNOS in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Canine, Drosophila, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen eNOS
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Constitutive NOS, EC 1.14.13.39, ECNOS, endothelial, Endothelial NOS, eNOScNOS, nitric oxide synthase 3 (endothelial cell), NOS type III, NOSIII
Gene Symbols NOS3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues S(1179) LQERQLRGAVPWAFD(1194) of human eNOS.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4846
Test Specificity This does not detect human inducible NOS or rat brain NOS.
Target Species Human, Mouse, Rat, Pig, Amphibian, Bovine, Canine, Drosophila, Sheep
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.