Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eNOS Antibody (M221), Novus Biologicals™
SDP

Catalog No. NB260182 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260182 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB260182 Supplier Novus Biologicals Supplier No. NBP323209
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

eNOS Monoclonal antibody specifically detects eNOS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence

Specifications

Antigen eNOS
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone M221
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50
Formulation PBS, 1 mg/ml BSA and 50% glycerol
Gene Alias Constitutive NOS, EC 1.14.13.39, ECNOS, endothelial, Endothelial NOS, eNOScNOS, nitric oxide synthase 3 (endothelial cell), NOS type III, NOSIII
Host Species Mouse
Immunogen Clone (M221) was generated from a recombinant mouse eNOS protein that included amino acids residues in the C-terminal region. This sequence is conserved in human and rat eNOS, and has low homology to other NOS family members. [UniProt# P70313]
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4846
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.