Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eNOS, p Ser116 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP229664
Description
eNOS Polyclonal specifically detects eNOS in Human, Bovine samples. It is validated for Western Blot.Specifications
eNOS (p Ser116) | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
P29474 | |
NOS3 | |
Peptide corresponding to amino acids 106-122 of human eNOS/NOS III (Ser116). | |
Affinity Purified | |
RUO | |
Primary | |
eNOS/NOS III phosphorylated on serine 116. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
0.2M Tris-Glycine (pH 7.4), 0.15M NaCl, 0.1mM EDTA and 30% Glycerol with 0.05% Sodium Azide | |
Constitutive NOS, EC 1.14.13.39, ECNOS, endothelial, Endothelial NOS, eNOScNOS, nitric oxide synthase 3 (endothelial cell), NOS type III, NOSIII | |
Rabbit | |
132 kDa | |
0.1 mL | |
Cancer, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, Phospho Specific | |
4846 | |
Human, Bovine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction