Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
eNOS, p Ser116 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$304.50 - $1073.00
Specifications
Antigen | eNOS (p Ser116) |
---|---|
Dilution | Western Blot 1:100-1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
eNOS Polyclonal specifically detects eNOS in Human, Bovine samples. It is validated for Western Blot.Specifications
eNOS (p Ser116) | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Bovine | |
P29474 | |
4846 | |
Peptide corresponding to amino acids 106-122 of human eNOS/NOS III (Ser116). | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
132 kDa |
Western Blot 1:100-1:2000 | |
Polyclonal | |
Rabbit | |
Cancer, HIF Target Genes, Hypoxia, Neuroscience, Neurotransmission, Phospho Specific | |
0.2M Tris-Glycine (pH 7.4), 0.15M NaCl, 0.1mM EDTA and 30% Glycerol with 0.05% Sodium Azide | |
Constitutive NOS, EC 1.14.13.39, ECNOS, endothelial, Endothelial NOS, eNOScNOS, nitric oxide synthase 3 (endothelial cell), NOS type III, NOSIII | |
NOS3 | |
IgG | |
Affinity Purified | |
eNOS/NOS III phosphorylated on serine 116. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title