Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENPP-1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB338673 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338673 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338673 Supplier Novus Biologicals Supplier No. NB600816APC
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ENPP-1 Polyclonal antibody specifically detects ENPP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA

Specifications

Antigen ENPP-1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA
Formulation PBS
Gene Alias ectonucleotide pyrophosphatase/phosphodiesterase 1, E-NPP 1, Ly-41 antigen, M6S1NPP1, Membrane component chromosome 6 surface marker 1, membrane component, chromosome 6, surface marker 1, NPPSalkaline phosphodiesterase 1, PC1, PC-1, PCA1ARHR2, PDNP1ectonucleotide pyrophosphatase/phosphodiesterase family member 1, Phosphodiesterase I/nucleotide pyrophosphatase 1, plasma-cell membrane glycoprotein 1, Plasma-cell membrane glycoprotein PC-1
Host Species Goat
Immunogen A synthetic peptide made to a C-terminal portion of the human ENPP1 protein (between residues 900-925) [UniProt P22413]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5167
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.