Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENPP-1 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB600816UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600816UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600816UV Supplier Novus Biologicals Supplier No. NB600816UV
Add to Cart
Add to Cart

Goat Polyclonal Antibody

ENPP-1 Polyclonal specifically detects ENPP-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen ENPP-1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ectonucleotide pyrophosphatase/phosphodiesterase 1, E-NPP 1, Ly-41 antigen, M6S1NPP1, Membrane component chromosome 6 surface marker 1, membrane component, chromosome 6, surface marker 1, NPPSalkaline phosphodiesterase 1, PC1, PC-1, PCA1ARHR2, PDNP1ectonucleotide pyrophosphatase/phosphodiesterase family member 1, Phosphodiesterase I/nucleotide pyrophosphatase 1, plasma-cell membrane glycoprotein 1, Plasma-cell membrane glycoprotein PC-1
Gene Symbols ENPP1
Host Species Goat
Immunogen A synthetic peptide made to a C-terminal portion of the human ENPP1 protein (between residues 900-925) [UniProt P22413]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5167
Target Species Human
Content And Storage Store at 4°C in the dark.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.