Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENPP-3/CD203c Antibody (NP4D6), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB296046 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB296046 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB296046 Supplier Novus Biologicals Supplier No. FAB5756MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ENPP-3/CD203c Monoclonal antibody specifically detects ENPP-3/CD203c in Human samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen ENPP-3/CD203c
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone NP4D6
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD203c, CD203c antigen, dJ1005H11.3 (phosphodiesterase I/nucleotide pyrophosphatase 3), dJ914N13.3 (phosphodiesterase I/nucleotide pyrophosphatase 3), ectonucleotide pyrophosphatase/phosphodiesterase 3, ectonucleotide pyrophosphatase/phosphodiesterase family member 3, E-NPP 3, gp130RB13-6, NPP3, PD-IBETA, PDNP3B10, Phosphodiesterase I beta, Phosphodiesterase I/nucleotide pyrophosphatase 3, phosphodiesterase-I beta
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human ENPP-3/CD203c, Accession # O14638
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Phosphatase
Primary or Secondary Primary
Gene ID (Entrez) 5169
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.