Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ENT1 Rabbit anti-Human, Alexa Fluor 488, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB045028 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB045028 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB045028 Supplier Novus Biologicals Supplier No. NBP297568AF488

Rabbit Polyclonal Antibody

ENT1 Polyclonal specifically detects ENT1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen ENT1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ENT1es-type, equilibrative nucleoside transporter 1, MGC1465, MGC3778, solute carrier family 29 (nucleoside transporters), member 1, Solute carrier family 29 member 1, solute carrier family 29, member 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ENT1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2030
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.