Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Entacapone
GREENER_CHOICE

Catalog No. p-4816692
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent COMT inhibitor; blocks α-synuclein aggregation

Potent catechol O-methyltransferase (COMT) inhibitor (IC50 values are 14.3, 20.1 and 73.3 nM for rat liver soluble COMT, total COMT and membrane-bound COMT respectively). Increases bioavailability of levodopa when given as an adjunct therapy for Parkinson's disease. Inhibits α-synuclein aggregation in an in vitro assay; blocks α-synuclein-induced cell death in PC-12 cells.

Chemical Identifiers

CAS 130929-57-6
Molecular Formula C14H15N3O5

Specifications

Quantity 10 mg
Formula Weight 305.29
Purity >99%
Molecular Weight (g/mol) 305.29
Chemical Name or Material Entacapone
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.