Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enterobacter aerogenes Antibody (57/82) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB119595 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119595 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119595 Supplier Novus Biologicals Supplier No. NBP308869

Mouse Monoclonal Antibody

Enterobacter aerogenes Monoclonal specifically detects Enterobacter aerogenes in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Enterobacter aerogenes
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 57/82
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA : 1-5 ug/mL, Immunocytochemistry/Immunofluorescence : 1-2 ug/mL
Formulation 10mM PBS
Host Species Mouse
Immunogen A crude sonicate of Enterobacter aerogenes
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.