Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Enterobacter aerogenes Mouse anti-Bacteria, Clone: 57/82, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30754520UG
Description
Enterobacter aerogenes Monoclonal specifically detects Enterobacter aerogenes in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Enterobacter aerogenes | |
Monoclonal | |
0.2 mg/ml | |
ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml | |
Mouse | |
Protein A or G purified | |
RUO | |
Bacteria | |
Purified |
ELISA, Immunofluorescence | |
57/82 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
A crude sonicate of Enterobacter aerogenes | |
20 μg | |
Primary | |
Store at 4°C. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction