Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Enterobacter aerogenes Mouse anti-Bacteria, Clone: 57/82, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | Enterobacter aerogenes |
---|---|
Clone | 57/82 |
Concentration | 0.2 mg/ml |
Dilution | ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml |
Applications | ELISA, Immunofluorescence |
Description
Enterobacter aerogenes Monoclonal specifically detects Enterobacter aerogenes in Bacteria samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Enterobacter aerogenes | |
0.2 mg/ml | |
ELISA, Immunofluorescence | |
Unconjugated | |
Mouse | |
Bacteria | |
A crude sonicate of Enterobacter aerogenes | |
Primary | |
Store at 4°C. |
57/82 | |
ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
IgG κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title