Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enterobacter Cloacae Antibody (61/212), FITC, Novus Biologicals™
SDP

Catalog No. NB10062544F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062544F 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062544F Supplier Novus Biologicals Supplier No. NB10062544F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Enterobacter Cloacae Monoclonal specifically detects Enterobacter Cloacae in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Enterobacter Cloacae
Applications ELISA
Classification Monoclonal
Clone 61/212
Conjugate FITC
Dilution ELISA
Formulation PBS with 0.05% Sodium Azide
Gene Alias Bacillus cloacae, Bacterium cloacae, Cloaca cloacae
Host Species Mouse
Immunogen Enterobacter cloacae NCTC 10005
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This product recognizes Enterobacter cloacae, a gram-negative bacillus sometimes associated with urinary and respiratory tract infections in humans.
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.