Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enteropeptidase/Enterokinase Antibody (MM0259- 8S12), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP212411C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212411C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212411C Supplier Novus Biologicals Supplier No. NBP212411C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Enteropeptidase/Enterokinase Monoclonal specifically detects Enteropeptidase/Enterokinase in Human samples. It is validated for Western Blot.

Specifications

Antigen Enteropeptidase/Enterokinase
Applications Western Blot
Classification Monoclonal
Clone MM0259- 8S12
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.4.21, EC 3.4.21.9, Enterokinase, ENTKenterokinase, MGC133046, protease, serine, 7 (enterokinase), PRSS7enteropeptidase, Serine protease 7, Transmembrane protease serine 15, transmembrane protease, serine 15
Gene Symbols TMPRSS15
Host Species Mouse
Immunogen Human recombinant Enteropeptidase/Enterokinase
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline ECS Epitope Tags (DDDDK), Epitope Tags, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 5651
Test Specificity Detects human Enteropeptidase/Enterokinase heavy chain
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.