Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enteropeptidase/Enterokinase Antibody (MM0259- 8S12), PerCP, Novus Biologicals™
SDP

Catalog No. NB360957 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360957 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360957 Supplier Novus Biologicals Supplier No. NBP212411PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Enteropeptidase/Enterokinase Monoclonal antibody specifically detects Enteropeptidase/Enterokinase in Human samples. It is validated for Western Blot

Specifications

Antigen Enteropeptidase/Enterokinase
Applications Western Blot
Classification Monoclonal
Clone MM0259- 8S12
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias EC 3.4.21, EC 3.4.21.9, Enterokinase, ENTKenterokinase, MGC133046, protease, serine, 7 (enterokinase), PRSS7enteropeptidase, Serine protease 7, Transmembrane protease serine 15, transmembrane protease, serine 15
Host Species Mouse
Immunogen Human recombinant Enteropeptidase/Enterokinase
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epitope Tags, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 5651
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.