Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Enterovirus 71 3D Rabbit anti-Virus, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB167225 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167225 100 μg
NB167226 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167225 Supplier Novus Biologicals Supplier No. NBP316065100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Enterovirus 71 3D Polyclonal antibody specifically detects Enterovirus 71 3D in Virus samples. It is validated for Western Blot

Specifications

Antigen Enterovirus 71 3D
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias 3D, 3D polymerase, 3Dpol, EV71 3D, Protein 3D, RdRp, RNA-directed RNA polymerase
Host Species Rabbit
Immunogen Recombinant protein of Enterovirus Enterovirus 71 3D
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.