Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EOMES Antibody (1219A), HRP, Novus Biologicals™
SDP

Catalog No. NB345953 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345953 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345953 Supplier Novus Biologicals Supplier No. FAB8889H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

EOMES Monoclonal specifically detects EOMES in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen EOMES
Applications Flow Cytometry
Classification Monoclonal
Clone 1219A
Conjugate HRP
Dilution Flow Cytometry
Formulation PBS
Gene Alias eomesodermin, eomesodermin (Xenopus laevis) homolog, t box, brain, 2, T-box brain protein 2, T-box brain2, TBR-2, TBR2eomesodermin homolog, T-brain-2
Host Species Rabbit
Immunogen E. coli-derived recombinant mouse EOMES, Met1-Ser126, Accession # O54839
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 8320
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.