Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EOMES Antibody (644730), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB376953 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB376953 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB376953 Supplier Novus Biologicals Supplier No. FAB6166K
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EOMES Monoclonal antibody specifically detects EOMES in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF

Specifications

Antigen EOMES
Applications Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 644730
Conjugate DyLight 488
Dilution Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias eomesodermin, eomesodermin (Xenopus laevis) homolog, t box, brain, 2, T-box brain protein 2, T-box brain2, TBR-2, TBR2eomesodermin homolog, T-brain-2
Host Species Mouse
Immunogen E. coli-derived recombinant human EOMES, Gly471-Pro686, Accession # O95936
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 8320
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.