Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ EOMES Recombinant Protein Antigen
Shop All R&D Systems Products

Click to view available options
Quantity:
0.1 mL
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human EOMES. Source: E.coli Amino Acid Sequence: KGFRDNYDSMYTASENDRLTPSPTDSPRSHQIVPGGRYGVQSFFPEPFVNTLPQARYYNGERTVPQTNGLLSPQQSEEVANPPQRWLVTPVQQPGTNKLDISSYESEYTSSTLLPYGIKSLPLQTSHALGYYPDPTFPAMAGWGGRGSYQRKMAAGLPWTSRTSPTV The EOMES Recombinant Protein Antigen is derived from E. coli. The EOMES Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
This is a blocking peptide for NBP1-80699. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.
Specifications
Specifications
| Gene ID (Entrez) | 8320 |
| Species | Human |
| Purification Method | Chromatography |
| Purity | >80% |
| Concentration | 0.5mg/mL |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Formulation | PBS and 1M Urea, pH 7.4. |
| For Use With (Application) | Blocking/Neutralizing, Control |
| Gene Symbol | EOMES |
| Label Type | Unlabeled |
| Show More |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction