Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eosinophil Antibody (BMK-13), Novus Biologicals™
SDP

Catalog No. NBP1421401M Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1421401M 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP1421401M Supplier Novus Biologicals Supplier No. NBP1421401ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 6 publications

Eosinophil Monoclonal specifically detects Eosinophil in Human, Mouse, Canine, Monkey, Rhesus Macaque samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Eosinophil
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone BMK-13
Concentration 0.1 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Gene Alias BMPG, Bone marrow proteoglycan, EMBP, Eosinophil granule major basic protein, Eosinophil major basic protein, MBP, MGC14537, MPG, Natural killer cell activator, Pregnancy associated major basic protein, PRG 2, Proteoglycan 2, Proteoglycan 2 bone marrow, Proteoglycan 2 preproprotein, Proteoglycan2
Host Species Mouse
Immunogen The antibody reacts with the Eosinophil Major Basic Protein antigen.
Purification Method Protein A or G purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity BMK-13 binds to the 10 kD eosinophil Major Basic Protein (MBP) of both resting and activated eosinophils in cytospins and frozen sections of bronchial and skin biopsies of allergic sites and normal sites and thus can be used as a “pan-eosinophil” marker. The antibody BMK-13 stains in frozen sections of bronchial biopsies from atopic asthmatics, rhinitics and normal non-atopic subjects, substantially higher counts of positive cells when compared to EG1, EG2 and chromotrope 2R. BMK-13 cross-reacts weakly with human basophils, which also contain low level of this protein. It does not cross-react with any other human protein or cell.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.