Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epac2 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB185009 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB185009 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB185009 Supplier Novus Biologicals Supplier No. NBP282089AF700
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Epac2 Polyclonal antibody specifically detects Epac2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Epac2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias CAMP-GEFII, cAMP-regulated guanine nucleotide exchange factor II, CGEF2EPAC 2, EPAC2, Exchange factor directly activated by cAMP 2, Exchange protein directly activated by cAMP 2, Nbla00496, putative protein product of Nbla00496, Rap guanine nucleotide exchange factor (GEF) 4, rap guanine nucleotide exchange factor 4, RAP guanine-nucleotide-exchange factor (GEF) 4
Host Species Rabbit
Immunogen EPAC2 antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human EPAC2. The immunogen is located within the first 50 amino acids of EPAC2. Amino Acid Squence: LDKRPLERSSEDVD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11069
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.