Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPB41L4A Antibody, Novus Biologicals™
SDP

Catalog No. NB10093422 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093422 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10093422 Supplier Novus Biologicals Supplier No. NB10093422
Add to Cart
Add to Cart

Goat Polyclonal Antibody

EPB41L4A Polyclonal specifically detects EPB41L4A in Human, Mouse samples. It is validated for Peptide ELISA.

Specifications

Antigen EPB41L4A
Applications Peptide ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_071423.3
Gene Alias DKFZp566L203, EPB41L4erythrocyte protein band 4.1-like 4, erythrocyte membrane protein band 4.1 like 4A, FLJ38738, NBL4band 4.1-like protein 4A, Protein NBL4
Gene Symbols EPB41L4A
Host Species Goat
Immunogen Peptide with sequence C-KNENSPDTQRSKSH corresponding to internal region according to NP_071423.3.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64097
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.