Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EpCAM/TROP1 Antibody (002), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB099537 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB099537 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB099537 Supplier Novus Biologicals Supplier No. NBP289589JF549
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

EpCAM/TROP1 Monoclonal specifically detects EpCAM/TROP1 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA Detection.

Specifications

Antigen EpCAM/TROP1
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2
Conjugate Janelia Fluor 549
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA Detection
Formulation 50 mM Sodium Borate
Gene Alias 17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2EGP34, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EpCAM/TROP1 (NP_032558.2; Met 1-Thr 266).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cancer Stem Cells, Tumor Biomarkers
Primary or Secondary Primary
Gene ID (Entrez) 4072
Test Specificity No cross-reactivity in ELISA with: Human EPCAM
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.