Learn More
Description
Specifications
Specifications
| Antigen | 58K Golgi Protein |
| Applications | Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 58K-9 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:2000 - 1:5000, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:250, Immunohistochemistry-Paraffin 1:100 - 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 24505439) |
| Formulation | Ascites with 0.1% Sodium Azide |
| Gene Accession No. | O95954, Q91XD4, O88618 |
| Gene Alias | formimidoyltransferase cyclodeaminase, formimidoyltransferase-cyclodeaminase, formiminotransferase cyclodeaminase, Formiminotransferase-cyclodeaminase, human formiminotransferase cyclodeaminase, EC 4.3.1.410formiminotransferase-cyclodeaminase, LCHC1 |
| Gene Symbols | FTCD |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
