Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

58K Golgi Protein Antibody (58K-9), Novus Biologicals™
SDP

Catalog No. NB600412SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600412SS 0.025 mL
NB600412 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB600412SS Supplier Novus Biologicals Supplier No. NB600412SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

58K Golgi Protein Monoclonal specifically detects 58K Golgi Protein in Bovine, Canine, Chicken, Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen 58K Golgi Protein
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 58K-9
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:5000, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:250, Immunohistochemistry-Paraffin 1:100 - 1:200, Immunohistochemistry-Frozen reported in scientific literature (PMID 24505439)
Formulation Ascites with 0.1% Sodium Azide
Gene Accession No. O95954, Q91XD4, O88618
Gene Alias formimidoyltransferase cyclodeaminase, formimidoyltransferase-cyclodeaminase, formiminotransferase cyclodeaminase, Formiminotransferase-cyclodeaminase, human formiminotransferase cyclodeaminase, EC 4.3.1.410formiminotransferase-cyclodeaminase, LCHC1
Gene Symbols FTCD
Host Species Mouse
Immunogen 58K Golgi Protein purified from rat liver [UniProt# O88618]
Molecular Weight of Antigen 58 kDa
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Golgi Apparatus Markers, Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10841
Test Specificity Rat's 58K Golgi Protein sequence is 92% similar with Chinese hamster (Cricetulus griseus).
Target Species Bovine, Canine, Chicken, Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.