Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EpCAM/TROP1 Antibody (EGP40/1798) - (ECD), Novus Biologicals™
SDP

Catalog No. NB25321320 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB25321320 20 μg
NB253213100 100 μg
NB253213200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB25321320 Supplier Novus Biologicals Supplier No. NBP25321320UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EpCAM/TROP1 Monoclonal specifically detects EpCAM/TROP1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array.

Specifications

Antigen EpCAM/TROP1
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EGP40/1798
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, ELISA 2 - 4 ug/ml, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500, SDS-Page
Formulation 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Gene Alias 17-1A, 323/A3, ACSTD1, antigen identified by monoclonal AUA1, CD326 antigen, Cell surface glycoprotein Trop-1, chromosome 4, surface marker (35kD glycoprotein), DIAR5, EGP, EGP-2, EGP314, EGP40, EpCAM, epithelial cell adhesion molecule, Epithelial cell surface antigen, Epithelial glycoprotein, Epithelial glycoprotein 314, ESA, GA733-2EGP34, hEGP314, HNPCC8, KS 1/4 antigen, KS1/4, KSAHEA125, M1S2, M4S1Ly74, Major gastrointestinal tumor-associated protein GA733-2, MIC18MH99, MOC31, TACST-1, TACSTD1, TROP1CD326, Tumor-associated calcium signal transducer 1CO-17A
Gene Symbols EPCAM
Host Species Mouse
Immunogen Recombinant human EpCAM protein fragment from the extracellular domain (aa77-202) (exact sequence is proprietary)
Molecular Weight of Antigen 42 kDa
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Cancer Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 4072
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.