Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPCR Antibody (016), DyLight 405
SDP

Catalog No. NB077184 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB077184 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB077184 Supplier Novus Biologicals Supplier No. NBP290739V

Rabbit Monoclonal Antibody

EPCR Monoclonal specifically detects EPCR in Mouse samples. It is validated for Flow Cytometry, ELISA.

Specifications

Antigen EPCR
Applications Flow Cytometry, ELISA
Classification Monoclonal
Clone 16
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias APC receptor, CCCA, CCD41centrosome-associated protein, CD201 antigen, centrocyclin, Endothelial cell protein C receptor, endothelial protein C receptor, EPCRMGC23024, protein C receptor, endothelial
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse EPCR (Q64695; Met1-Ser214).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 10544
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.