Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPCR Antibody (432714), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB313296 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313296 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313296 Supplier Novus Biologicals Supplier No. FAB2749APCCY7
Add to Cart
Add to Cart

Rat Monoclonal Antibody

EPCR Monoclonal specifically detects EPCR in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen EPCR
Applications Flow Cytometry
Classification Monoclonal
Clone 432714
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias APC receptor, CCCA, CCD41centrosome-associated protein, CD201 antigen, centrocyclin, Endothelial cell protein C receptor, endothelial protein C receptor, EPCRMGC23024, protein C receptor, endothelial
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse EPCR, Ala17-Ser214, Accession # Q64695
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 10544
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.