Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPCR Antibody (RCR-252) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB600963SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600963SS 0.025 mL
NB600963 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB600963SS Supplier Novus Biologicals Supplier No. NB600963SS
Add to Cart
Add to Cart

Rat Monoclonal Antibody

EPCR Monoclonal specifically detects EPCR in Human samples. It is validated for Flow Cytometry, Microarray, Block/Neutralize.

Specifications

Antigen EPCR
Applications Flow Cytometry, Blocking Assay
Classification Monoclonal
Clone RCR-252
Concentration 2.4 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 5-20 ug/ml, Blocking/Neutralizing 50-200 molar excess, Microarray
Formulation 10mM PBS (pH 7.4) with 15mM Sodium Azide
Gene Alias APC receptor, CCCA, CCD41centrosome-associated protein, CD201 antigen, centrocyclin, Endothelial cell protein C receptor, endothelial protein C receptor, EPCRMGC23024, protein C receptor, endothelial
Gene Symbols PROCR
Host Species Rat
Immunogen Myeloma cells and cells isolated from the superficial inguinal lymph nodes from Wistar rats immunized with the human EPCR-positive RE-1 cells.
Molecular Weight of Antigen 49 kDa
Purification Method Protein A or G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 10544
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.