Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPDR1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB329454 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB329454 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB329454 Supplier Novus Biologicals Supplier No. NBP300177PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EPDR1 Polyclonal antibody specifically detects EPDR1 in Human samples. It is validated for Western Blot, ELISA

Specifications

Antigen EPDR1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate PE
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias EPDR, ependymin related protein 1 (zebrafish), MERP1mammalian ependymin related protein 1, MERP-1Upregulated in colorectal cancer gene 1 protein, UCC1mammalian ependymin-related protein 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human EPDR1 (Uniprot#: Q9UM22-1; Met1-Trp224)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54749
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.