Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EPDR1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB169105 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB169105 25 μg
NB169104 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB169105 Supplier Novus Biologicals Supplier No. NBP31701425UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EPDR1 Polyclonal antibody specifically detects EPDR1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen EPDR1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias EPDR, ependymin related protein 1 (zebrafish), MERP1mammalian ependymin related protein 1, MERP-1Upregulated in colorectal cancer gene 1 protein, UCC1mammalian ependymin-related protein 1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: GRQVMYQQSSGRNSRALLSYDGLNQRVRVLDERKALIPCKRLFEYILLYKDGVMFQIDQATKQCSKMTLTQPWDPL
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54749
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.