Learn More
Description
Specifications
Specifications
| Antigen | EPGN |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | RM0228-2L47 |
| Conjugate | Alexa Fluor 594 |
| Dilution | Western Blot |
| Formulation | 50 mM sodium borate with 0.05% sodium azide |
| Gene Alias | epigen, epithelial mitogen homolog (mouse), PRO9904, UNQ3072/PRO9904 |
| Gene Symbols | EPGN |
| Host Species | Rat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
