Description
- For use with Multiphor II Flatbed Electrophoresis System
- EPH/IEF Electrode cathode and anode
Specifications
Specifications
| For Use With (Equipment) | Multiphor™ II Flatbed Electrophoresis System |
| Type | Cathode Electrode |