Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA10 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB358447 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB358447 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB358447 Supplier Novus Biologicals Supplier No. NBP176318MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EphA10 Polyclonal antibody specifically detects EphA10 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen EphA10
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.10.1, EPH receptor A10, EphA10s protein, ephrin type-A receptor 10, FLJ16103, FLJ33655, MGC43817
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide of near the amino terminus of human EphA10. The immunogen is located within amino acids 120 - 170 of EphA10. Amino Acid Squence: RPRLGGSRPRKIDT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 284656
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.