Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA2 Antibody (1C1), Biotin, Novus Biologicals™
SDP

Catalog No. NB015324 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015324 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB015324 Supplier Novus Biologicals Supplier No. NBP281069B
Add to Cart
Add to Cart

Human Monoclonal Antibody

EphA2 Monoclonal specifically detects EphA2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen EphA2
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 1C1
Conjugate Biotin
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK
Gene Symbols EPHA2
Host Species Human
Immunogen 1C1 was isolated after screening of a non-immunized human Fab phage library. Selection was based on the ability to bind soluble human recombinant EphA2 (residues 183-510). The Fab was subsequently converted to full length human IgG1.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 1969
Test Specificity 1C1 binds specifically to the amino-termianal LBD (ligand-binding domain) region of the EphA2 ectodomain (IgG1 KD ∽0.8 nM; Fab KD ∽140 nM) and not to other related human Eph receptors such as EphA1, EphA2, and EphA4, or any EphB receptor family members.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.