Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA2 Antibody (233720), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB311703 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311703 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311703 Supplier Novus Biologicals Supplier No. FAB639PEATT594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

EphA2 Monoclonal antibody specifically detects EphA2 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen EphA2
Applications Flow Cytometry
Classification Monoclonal
Clone 233720
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse EphA2, Ala22-Ala535 (predicted), Accession # AAA82113
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 1969
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.