Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA2 Antibody (371805), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB363465 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB363465 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB363465 Supplier Novus Biologicals Supplier No. FAB3035Y
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EphA2 Monoclonal antibody specifically detects EphA2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF, Gene Knock-Out

Specifications

Antigen EphA2
Applications Flow Cytometry, Immunocytochemistry, CyTOF, Gene Knock-Out
Classification Monoclonal
Clone 371805
Conjugate DyLight 680
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated
Formulation 50mM Sodium Borate
Gene Alias ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human EphA2, Gln25-Asn534, Accession # P29317
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 1969
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.