Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA2 Antibody (66G9), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB162198 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162198 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB162198 Supplier Novus Biologicals Supplier No. NBP259479AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EphA2 Monoclonal antibody specifically detects EphA2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen EphA2
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 66G9
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias ARCC2, EC 2.7.10, EC 2.7.10.1, ECKepithelial cell receptor protein tyrosine kinase, EPH receptor A2, EphA2, ephrin type-A receptor 2, Epithelial cell kinase, soluble EPHA2 variant 1, Tyrosine-protein kinase receptor ECK
Host Species Mouse
Immunogen Recombinant human EPHA2 (27-537aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 1969
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.