Learn More
EPHA2 Mouse anti-Human, Unconjugated, Clone: 1A9C3, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | EPHA2 |
---|---|
Clone | 1A9C3 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-9016
![]() ![]() |
Proteintech Group Inc
667361IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The EPHA2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EPHA2. This antibody recognizes human antigen. The EPHA2 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
EPHA2 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
1969 | |
Primary |
1A9C3 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Human | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.