Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA4 Antibody (11C13), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB193604549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB193604549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB193604549 Supplier Novus Biologicals Supplier No. NB11093604JF549
Add to Cart
Add to Cart

Rat Monoclonal Antibody

EphA4 Monoclonal specifically detects EphA4 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen EphA4
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 11C13
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.7.10, EC 2.7.10.1, EK8, EPH receptor A4, EphA4, EPH-like kinase 8, ephrin type-A receptor 4, HEK8, receptor protein-tyrosine kinase HEK8, SEK, TYRO1, TYRO1 protein tyrosine kinase, Tyrosine-protein kinase receptor SEK, Tyrosine-protein kinase TYRO1
Gene Symbols EPHA4
Host Species Rat
Immunogen Mouse EphA4 (extracellular domain)
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2043
Test Specificity In this format, this does not cross-react with recombinant mouse EphA2, EphA6, EphA7, EphA8 and EphB2, EphB3, EphB4 or EphB6.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.