Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA5 Antibody (86731), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB312040 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB312040 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB312040 Supplier Novus Biologicals Supplier No. FAB541APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EphA5 Monoclonal antibody specifically detects EphA5 in Human, Rat samples. It is validated for Flow Cytometry

Specifications

Antigen EphA5
Applications Flow Cytometry
Classification Monoclonal
Clone 86731
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias EC 2.7.10, EC 2.7.10.1, EHK-1, EHK1Hek7, EK7, EPH homology kinase 1, Eph homology kinase-1, EPH receptor A5, EphA5, EPH-like kinase 7, ephrin type-A receptor 5, hEK7, receptor protein-tyrosine kinase HEK7, TYRO4HEK7CEK7, tyrosine-protein kinase receptor EHK-1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant rat EphA5, Ser58-Gln573 (Asp170Glu), Accession # P54757
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Neuroscience, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2044
Target Species Human, Rat
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.