Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EphA6 Antibody (9J12), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBT93605647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBT93605647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBT93605647 Supplier Novus Biologicals Supplier No. NB11093605AF647
Add to Cart
Add to Cart

Rat Monoclonal Antibody

EphA6 Monoclonal specifically detects EphA6 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen EphA6
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 9J12
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp434C1418, EC 2.7.10, EC 2.7.10.1, EHK2, EHK-2, EPA6, EPH homology kinase 2, EPH receptor A6, ephrin receptor EphA6, ephrin type-A receptor 6, FLJ35246, PRO57066
Gene Symbols EPHA6
Host Species Rat
Immunogen Mouse EphA6 (extracellular domain)
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 285220
Test Specificity Does not cross-react with recombinant mouse EphA2, EphA3, EphA4, EphA7, EphA8 and EphB2, EphB3, EphB4 or EphB6.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.